Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACACTTGCTAAGTGTCAGGCA[C/T]AAGTCATAGTGAGACTGCCCTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP2K3 - mitogen-activated protein kinase kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316332.1 | Intron | NP_001303261.1 | ||||
NM_002756.4 | Intron | NP_002747.2 | ||||
NM_145109.2 | Intron | NP_659731.1 | ||||
XM_005256723.2 | Intron | XP_005256780.1 | ||||
XM_011523958.2 | Intron | XP_011522260.1 | ||||
XM_011523959.1 | Intron | XP_011522261.1 | ||||
XM_017024857.1 | Intron | XP_016880346.1 | ||||
XM_017024858.1 | Intron | XP_016880347.1 | ||||
XM_017024859.1 | Intron | XP_016880348.1 |