Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAAAACAAGCATTATGAGATGA[C/T]GTATATTTTGGAGAGATTATGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601545 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAFAH1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAFAH1B1 - platelet activating factor acetylhydrolase 1b regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000430.3 | Intron | NP_000421.1 | ||||
XM_011523901.1 | Intron | XP_011522203.1 | ||||
XM_011523902.2 | Intron | XP_011522204.1 | ||||
XM_011523903.2 | Intron | XP_011522205.1 | ||||
XM_017024699.1 | Intron | XP_016880188.1 | ||||
XM_017024700.1 | Intron | XP_016880189.1 | ||||
XM_017024701.1 | Intron | XP_016880190.1 | ||||
XM_017024702.1 | Intron | XP_016880191.1 | ||||
XM_017024703.1 | Intron | XP_016880192.1 |