Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGTCCAGGAGGCGGGTCAGACT[G/T]CCACAGAGGAGTTTGACCACGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611627 MIM: 606969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBIL5P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBIL5P - diazepam binding inhibitor-like 5, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM57A - family with sequence similarity 57 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GEMIN4 - gem nuclear organelle associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015721.2 | 3172 | Silent Mutation | GGA,GGC | G,G 945 | NP_056536.2 | |
XM_005256667.4 | 3172 | Silent Mutation | GGA,GGC | G,G 949 | XP_005256724.1 | |
XM_005256670.4 | 3172 | Silent Mutation | GGA,GGC | G,G 934 | XP_005256727.1 | |
XM_011523910.2 | 3172 | Silent Mutation | GGA,GGC | G,G 949 | XP_011522212.1 | |
XM_011523911.2 | 3172 | Silent Mutation | GGA,GGC | G,G 949 | XP_011522213.1 | |
XM_011523912.2 | 3172 | Silent Mutation | GGA,GGC | G,G 934 | XP_011522214.1 | |
XM_011523913.2 | 3172 | Silent Mutation | GGA,GGC | G,G 934 | XP_011522215.1 | |
XM_017024709.1 | 3172 | Silent Mutation | GGA,GGC | G,G 949 | XP_016880198.1 |