Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTTTCTCCAGCTGGGACTCTAG[C/T]GACTTCATCTTTCTTCCCACATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASGR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASGR1 - asialoglycoprotein receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197216.2 | 619 | Silent Mutation | TCA,TCG | S,S 65 | NP_001184145.1 | |
NM_001671.4 | 619 | Silent Mutation | TCA,TCG | S,S 104 | NP_001662.1 | |
XM_011523861.2 | 619 | Silent Mutation | TCA,TCG | S,S 104 | XP_011522163.1 |