Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCTGGAGCCCCAGTCCTAAGAA[G/T]AAAAAGCAGGCCACGGGGTTAGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLG4 - discs large MAGUK scaffold protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128827.2 | Intron | NP_001122299.1 | ||||
NM_001321074.1 | Intron | NP_001308003.1 | ||||
NM_001321075.1 | Intron | NP_001308004.1 | ||||
NM_001321076.1 | Intron | NP_001308005.1 | ||||
NM_001321077.1 | Intron | NP_001308006.1 | ||||
NM_001365.4 | Intron | NP_001356.1 | ||||
XM_005256491.1 | Intron | XP_005256548.1 | ||||
XM_011523698.1 | Intron | XP_011522000.1 | ||||
XM_011523699.2 | Intron | XP_011522001.1 | ||||
XM_011523702.1 | Intron | XP_011522004.1 | ||||
XM_017024288.1 | Intron | XP_016879777.1 | ||||
XM_017024289.1 | Intron | XP_016879778.1 | ||||
XM_017024290.1 | Intron | XP_016879779.1 |