Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTGCTGCTTGAGGATTCACTGC[A/G]TAGCCAAAGACCCCTGGTAGGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100710 MIM: 612308 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNB1 - cholinergic receptor nicotinic beta 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB4 - zinc finger and BTB domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128833.1 | 3519 | Silent Mutation | TAC,TAT | Y,Y 970 | NP_001122305.1 | |
NM_020899.3 | 3519 | Silent Mutation | TAC,TAT | Y,Y 970 | NP_065950.2 | |
XM_006721563.3 | 3519 | Silent Mutation | TAC,TAT | Y,Y 970 | XP_006721626.1 | |
XM_006721564.2 | 3519 | Silent Mutation | TAC,TAT | Y,Y 970 | XP_006721627.1 | |
XM_011523972.2 | 3519 | Silent Mutation | TAC,TAT | Y,Y 970 | XP_011522274.1 |