Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTTACCCACTGTGGTCCCTGG[A/G]TCGAGAGGTGACCACCAATCAGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602297 MIM: 612661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFNB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFNB3 - ephrin B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WRAP53 - WD repeat containing antisense to TP53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143990.1 | 1378 | Missense Mutation | GAT,GGT | D,G 408 | NP_001137462.1 | |
NM_001143991.1 | 1378 | Missense Mutation | GAT,GGT | D,G 408 | NP_001137463.1 | |
NM_001143992.1 | 1378 | Missense Mutation | GAT,GGT | D,G 408 | NP_001137464.1 | |
NM_018081.2 | 1378 | Missense Mutation | GAT,GGT | D,G 408 | NP_060551.2 | |
XM_011523952.2 | 1378 | Missense Mutation | GAT,GGT | D,G 195 | XP_011522254.1 | |
XM_017024812.1 | 1378 | Intron | XP_016880301.1 |