Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCACTCGTAGTCGGGAAGAGGAG[A/C]TTCAGCAGCGCTGTTGTCCCACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611425 MIM: 604111 MIM: 610969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTROB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNAB3 - potassium voltage-gated channel subfamily A regulatory beta subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC1 - trafficking protein particle complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166621.1 | 529 | Intron | NP_001160093.1 | |||
NM_021210.4 | 529 | Intron | NP_067033.1 |