Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGCTCTGTCCATCACCAGAGCA[C/T]ACCTGCCCAGGCTGGCACACCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602260 MIM: 185881 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PER1 - period circadian clock 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAMP2 - vesicle associated membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014232.2 | Intron | NP_055047.2 | ||||
XM_005256775.4 | Intron | XP_005256832.1 |