Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGCATCCCTCAAGCCCCCTGCT[A/C]TTTTGCCCCCCTCAGCTTCTAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608504 | ||||||||||||||||||||
Literature Links: |
ARHGEF15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF15 - Rho guanine nucleotide exchange factor 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025014.1 | 314 | Missense Mutation | ATT,CTT | I,L 75 | NP_079290.1 | |
NM_173728.3 | 314 | Missense Mutation | ATT,CTT | I,L 75 | NP_776089.2 | |
XM_011523734.1 | 314 | Missense Mutation | ATT,CTT | I,L 75 | XP_011522036.1 | |
XM_011523735.1 | 314 | Missense Mutation | ATT,CTT | I,L 75 | XP_011522037.1 | |
XM_011523736.1 | 314 | Missense Mutation | ATT,CTT | I,L 75 | XP_011522038.1 | |
XM_011523737.1 | 314 | Intron | XP_011522039.1 |