Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTCCCCTCAATGAATTCTTTC[C/T]CTGTAAGTTTATCTGTGCATTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603659 MIM: 179618 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLP2R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLP2R - glucagon like peptide 2 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCVRN - recoverin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002903.2 | 693 | Missense Mutation | GAG,GGG | E,G 169 | NP_002894.1 |