Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCCCACCGCCAGCCCCCGCCCC[G/T]CCCTGCACTGATTTAGACCTGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608476 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBKBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBKBP1 - TBK1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014726.2 | 1422 | Silent Mutation | CCG,CCT | P,P 191 | NP_055541.1 | |
XM_005257859.4 | 1422 | Silent Mutation | CCG,CCT | P,P 191 | XP_005257916.1 | |
XM_005257860.4 | 1422 | Silent Mutation | CCG,CCT | P,P 191 | XP_005257917.1 | |
XM_005257861.4 | 1422 | Silent Mutation | CCG,CCT | P,P 191 | XP_005257918.1 | |
XM_005257862.3 | 1422 | Silent Mutation | CCG,CCT | P,P 191 | XP_005257919.1 | |
XM_017025431.1 | 1422 | Silent Mutation | CCG,CCT | P,P 191 | XP_016880920.1 | |
XM_017025432.1 | 1422 | Silent Mutation | CCG,CCT | P,P 189 | XP_016880921.1 |