Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTGGTCTCAGCCGGACTCGGGT[A/T]TCCGGGCAGGGTGGGACCAGCCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615526 MIM: 163260 | ||||||||||||||||||||
Literature Links: |
COPZ2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPZ2 - coatomer protein complex subunit zeta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016429.3 | Intron | NP_057513.1 | ||||
XM_011524874.2 | Intron | XP_011523176.1 |
NFE2L1 - nuclear factor, erythroid 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003204.2 | Intron | NP_003195.1 | ||||
XM_005257410.3 | Intron | XP_005257467.1 | ||||
XM_005257411.3 | Intron | XP_005257468.1 | ||||
XM_005257412.3 | Intron | XP_005257469.1 | ||||
XM_005257413.3 | Intron | XP_005257470.1 | ||||
XM_005257415.3 | Intron | XP_005257472.1 | ||||
XM_017024691.1 | Intron | XP_016880180.1 |