Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCCCTTTCTTTTTTACTGTTGC[A/G]TAATATTCACTTCTATGAATGTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SKAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SKAP1 - src kinase associated phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001075099.1 | Intron | NP_001068567.1 | ||||
NM_003726.3 | Intron | NP_003717.3 | ||||
XM_005257755.3 | Intron | XP_005257812.1 | ||||
XM_017025257.1 | Intron | XP_016880746.1 | ||||
XM_017025258.1 | Intron | XP_016880747.1 | ||||
XM_017025259.1 | Intron | XP_016880748.1 | ||||
XM_017025260.1 | Intron | XP_016880749.1 |