Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCGCCTGCCTTGCTTCTCTTTC[C/T]AGCCTTCCTACAGCAACTTCCCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603192 | ||||||||||||||||||||
Literature Links: |
ATP5G1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5G1 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002027.1 | Intron | NP_001002027.1 | ||||
NM_005175.2 | Intron | NP_005166.1 |
LOC105371814 - uncharacterized LOC105371814 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |