Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCACGTGGCTTCTTCAAACACA[C/T]GCACTTTCTCACAGCCTTCTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM117A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM117A - family with sequence similarity 117 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030802.3 | 1870 | Missense Mutation | CAT,CGT | H,R 348 | NP_110429.1 | |
XM_017025182.1 | 1870 | Missense Mutation | CAT,CGT | H,R 396 | XP_016880671.1 |
SLC35B1 - solute carrier family 35 member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |