Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGTAATGTCAACCTTTACTTTA[C/T]GGTATCTTAGGTTTGTCTTCTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAG9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPAG9 - sperm associated antigen 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130527.2 | 5366 | UTR 3 | NP_001123999.1 | |||
NM_001130528.2 | 5366 | UTR 3 | NP_001124000.1 | |||
NM_001251971.1 | 5366 | UTR 3 | NP_001238900.1 | |||
NM_003971.5 | 5366 | UTR 3 | NP_003962.3 | |||
XM_005257768.2 | 5366 | UTR 3 | XP_005257825.1 | |||
XM_005257771.2 | 5366 | UTR 3 | XP_005257828.1 | |||
XM_005257774.3 | 5366 | UTR 3 | XP_005257831.1 | |||
XM_017025283.1 | 5366 | UTR 3 | XP_016880772.1 | |||
XM_017025284.1 | 5366 | UTR 3 | XP_016880773.1 | |||
XM_017025285.1 | 5366 | UTR 3 | XP_016880774.1 |