Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAATGTCTAGTGTGCCAGGCCT[A/T]TCTTGATTTTTCTCATCTTTTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616334 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM100 - transmembrane protein 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099640.1 | 2899 | UTR 3 | NP_001093110.1 | |||
NM_018286.2 | 2899 | UTR 3 | NP_060756.2 | |||
XM_017024814.1 | 2899 | UTR 3 | XP_016880303.1 | |||
XM_017024815.1 | 2899 | UTR 3 | XP_016880304.1 | |||
XM_017024816.1 | 2899 | UTR 3 | XP_016880305.1 |