Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTCTCTTGAATTACTTGAGTGA[C/T]TTGAATTCTATTAAAAGCACCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP1 - A-kinase anchoring protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242902.1 | Intron | NP_001229831.1 | ||||
NM_001242903.1 | Intron | NP_001229832.1 | ||||
NM_003488.3 | Intron | NP_003479.1 | ||||
XM_005257707.2 | Intron | XP_005257764.1 | ||||
XM_005257709.2 | Intron | XP_005257766.1 | ||||
XM_017025189.1 | Intron | XP_016880678.1 | ||||
XM_017025190.1 | Intron | XP_016880679.1 | ||||
XM_017025191.1 | Intron | XP_016880680.1 |