Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCCCCACGTTCAATCCTGGGG[A/G]CGCTGCCAGAGCTGGGGGTACAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610764 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPOAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPOAP1 - TSPO associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261835.1 | 5532 | UTR 3 | NP_001248764.1 | |||
NM_004758.3 | 5532 | UTR 3 | NP_004749.2 | |||
NM_024418.2 | 5532 | UTR 3 | NP_077729.1 | |||
XM_005257807.2 | 5532 | Missense Mutation | GCC,GTC | A,V 1863 | XP_005257864.1 | |
XM_006722173.2 | 5532 | Missense Mutation | GCC,GTC | A,V 1872 | XP_006722236.1 | |
XM_006722174.2 | 5532 | Missense Mutation | GCC,GTC | A,V 1865 | XP_006722237.1 | |
XM_006722175.2 | 5532 | Missense Mutation | GCC,GTC | A,V 1812 | XP_006722238.1 | |
XM_006722176.2 | 5532 | Missense Mutation | GCC,GTC | A,V 1595 | XP_006722239.1 | |
XM_006722177.3 | 5532 | Missense Mutation | GCC,GTC | A,V 1432 | XP_006722240.1 | |
XM_006722178.3 | 5532 | Missense Mutation | GCC,GTC | A,V 1432 | XP_006722241.1 | |
XM_011525471.2 | 5532 | Missense Mutation | GCC,GTC | A,V 1393 | XP_011523773.1 | |
XM_011525472.1 | 5532 | Missense Mutation | GCC,GTC | A,V 1376 | XP_011523774.1 | |
XM_017025326.1 | 5532 | Intron | XP_016880815.1 | |||
XM_017025327.1 | 5532 | Missense Mutation | GCC,GTC | A,V 1833 | XP_016880816.1 | |
XM_017025328.1 | 5532 | Missense Mutation | GCC,GTC | A,V 1769 | XP_016880817.1 | |
XM_017025329.1 | 5532 | Missense Mutation | GCC,GTC | A,V 1592 | XP_016880818.1 | |
XM_017025330.1 | 5532 | UTR 3 | XP_016880819.1 | |||
XM_017025331.1 | 5532 | UTR 3 | XP_016880820.1 | |||
XM_017025332.1 | 5532 | UTR 3 | XP_016880821.1 | |||
XM_017025333.1 | 5532 | UTR 3 | XP_016880822.1 | |||
XM_017025334.1 | 5532 | UTR 3 | XP_016880823.1 |