Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTCTTCGTTTGGCCAACCAGAA[C/T]TCACAGTCACAGTTATAGCAGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603559 | ||||||||||||||||||||
Literature Links: |
HSF5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSF5 - heat shock transcription factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTMR4 - myotubularin related protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004687.4 | 3605 | Silent Mutation | GAA,GAG | E,E 1126 | NP_004678.3 | |
XM_005257784.2 | 3605 | Silent Mutation | GAA,GAG | E,E 1140 | XP_005257841.1 | |
XM_005257785.4 | 3605 | Silent Mutation | GAA,GAG | E,E 1130 | XP_005257842.1 | |
XM_005257786.4 | 3605 | Silent Mutation | GAA,GAG | E,E 1126 | XP_005257843.1 | |
XM_006722168.3 | 3605 | Silent Mutation | GAA,GAG | E,E 1126 | XP_006722231.1 | |
XM_011525460.2 | 3605 | Silent Mutation | GAA,GAG | E,E 1130 | XP_011523762.1 |