Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCAACACCGTGGATGAGCCCTG[A/C]TGCTGTTTCAAGAGCTCTGTCACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605792 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT4-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPT4-AS1 - SEPT4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX14 - testis expressed 14, intercellular bridge forming factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201457.1 | 4243 | Missense Mutation | CAG,CAT | Q,H 1347 | NP_001188386.1 | |
NM_031272.4 | 4243 | Missense Mutation | CAG,CAT | Q,H 1301 | NP_112562.3 | |
NM_198393.3 | 4243 | Missense Mutation | CAG,CAT | Q,H 1341 | NP_938207.2 | |
XM_011525029.2 | 4243 | Missense Mutation | CAG,CAT | Q,H 1382 | XP_011523331.1 | |
XM_017024860.1 | 4243 | Missense Mutation | CAG,CAT | Q,H 1160 | XP_016880349.1 | |
XM_017024861.1 | 4243 | Missense Mutation | CAG,CAT | Q,H 949 | XP_016880350.1 |