Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGCATAGACGTCAACAGAGGGG[C/T]CGAATTCCCCAGGACCAAAGAACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603261 MIM: 602176 | ||||||||||||||||||||
Literature Links: |
PIP4K2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIP4K2B - phosphatidylinositol-5-phosphate 4-kinase type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003559.4 | 1472 | Missense Mutation | GAC,GGC | D,G 343 | NP_003550.1 | |
XM_011525326.2 | 1472 | Missense Mutation | GAC,GGC | D,G 370 | XP_011523628.1 | |
XM_011525327.2 | 1472 | Missense Mutation | GAC,GGC | D,G 306 | XP_011523629.1 | |
XM_011525329.2 | 1472 | Intron | XP_011523631.1 | |||
XM_011525330.2 | 1472 | Missense Mutation | GAC,GGC | D,G 211 | XP_011523632.1 | |
XM_017025196.1 | 1472 | Missense Mutation | GAC,GGC | D,G 279 | XP_016880685.1 | |
XM_017025197.1 | 1472 | Missense Mutation | GAC,GGC | D,G 205 | XP_016880686.1 | |
XM_017025198.1 | 1472 | Missense Mutation | GAC,GGC | D,G 184 | XP_016880687.1 | |
XM_017025199.1 | 1472 | Missense Mutation | GAC,GGC | D,G 175 | XP_016880688.1 |
PSMB3 - proteasome subunit beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |