Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACACAATTCGCCTTCAAACTCCA[C/T]GACAAGGACCCTGAGAAGGCAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616159 MIM: 612275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHRS11 - dehydrogenase/reductase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024308.3 | 844 | Silent Mutation | CAC,CAT | H,H 210 | NP_077284.2 | |
XM_005257658.2 | 844 | Intron | XP_005257715.1 | |||
XM_011525233.2 | 844 | Silent Mutation | CAC,CAT | H,H 140 | XP_011523535.1 |
GGNBP2 - gametogenetin binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRM1 - mitochondrial rRNA methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |