Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTTTCCTGTGGGCTGTGGGGTG[C/T]TCCAGTGTCATCTCCCTCTGCTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HEATR6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEATR6 - HEAT repeat containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022070.4 | 5315 | Missense Mutation | ACA,GCA | T,A 1121 | NP_071353.4 | |
XM_005257570.3 | 5315 | Missense Mutation | ACA,GCA | T,A 953 | XP_005257627.1 | |
XM_011525110.2 | 5315 | Missense Mutation | ACA,GCA | T,A 1085 | XP_011523412.1 | |
XM_011525111.2 | 5315 | Missense Mutation | ACA,GCA | T,A 1074 | XP_011523413.1 | |
XM_017024936.1 | 5315 | Missense Mutation | ACA,GCA | T,A 953 | XP_016880425.1 | |
XM_017024937.1 | 5315 | Missense Mutation | ACA,GCA | T,A 953 | XP_016880426.1 | |
XM_017024938.1 | 5315 | Missense Mutation | ACA,GCA | T,A 660 | XP_016880427.1 | |
XM_017024939.1 | 5315 | Missense Mutation | ACA,GCA | T,A 660 | XP_016880428.1 |
MIR4737 - microRNA 4737 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |