Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGCATGTACAAGATGGAACAAA[C/T]TCAAAAAGCTTCTGTCATCTTCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609498 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO47 - F-box protein 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008777.2 | 1379 | Missense Mutation | AAT,AGT | N,S 426 | NP_001008777.2 | |
XM_011524865.2 | 1379 | Missense Mutation | AAT,AGT | N,S 400 | XP_011523167.1 | |
XM_011524866.2 | 1379 | Missense Mutation | AAT,AGT | N,S 369 | XP_011523168.1 | |
XM_011524867.2 | 1379 | Intron | XP_011523169.1 |
LINC00672 - long intergenic non-protein coding RNA 672 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |