Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTAAGTCATTTGTTTATTTTT[A/G]TCCTAGGAAATGCTGTATGTGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605324 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APPBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APPBP2 - amyloid beta precursor protein binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282476.1 | 3460 | UTR 3 | NP_001269405.1 | |||
NM_006380.3 | 3460 | UTR 3 | NP_006371.2 | |||
XM_011524198.2 | 3460 | UTR 3 | XP_011522500.1 | |||
XM_011524199.2 | 3460 | UTR 3 | XP_011522501.1 |