Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCACAGTTAGGCTGCTTGGTAG[G/T]ATCAGTGGAGTAGTCAAACTGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605973 MIM: 608168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF7 - DDB1 and CUL4 associated factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005828.4 | Intron | NP_005819.3 |
KCNH6 - potassium voltage-gated channel subfamily H member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |