Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTGTTCACCACGGCTTCTGCCT[C/T]CTTCCTGGTCTGCTCCAGCCTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613369 MIM: 601681 | ||||||||||||||||||||
Literature Links: |
DDX42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX42 - DEAD-box helicase 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007372.3 | 2933 | Intron | NP_031398.2 | |||
NM_203499.2 | 2933 | Intron | NP_987095.1 | |||
XM_006721657.1 | 2933 | Intron | XP_006721720.1 | |||
XM_011524259.2 | 2933 | Intron | XP_011522561.1 | |||
XM_017024111.1 | 2933 | Intron | XP_016879600.1 | |||
XM_017024112.1 | 2933 | Intron | XP_016879601.1 |
FTSJ3 - FtsJ homolog 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017647.3 | 2933 | Missense Mutation | AAG,GAG | K,E 763 | NP_060117.3 |
PSMC5 - proteasome 26S subunit, ATPase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |