Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAGACATCCTGTTTTAATTTTT[A/C]TTACTGAAAAAGTAAGTGATATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615366 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOL11 - nucleolar protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303272.1 | 623 | Intron | NP_001290201.1 | |||
NM_015462.4 | 623 | Missense Mutation | ATT,CTT | I,L 170 | NP_056277.2 |