Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCAGCACGATGTGGAGGAGACC[A/G]ATGGCCAGCACGTAGGAGAGCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611221 MIM: 610075 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSDMB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSDMB - gasdermin B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC3C - leucine rich repeat containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORMDL3 - ORMDL sphingolipid biosynthesis regulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320801.1 | 125 | Silent Mutation | ATC,ATT | I,I 30 | NP_001307730.1 | |
NM_001320802.1 | 125 | Silent Mutation | ATC,ATT | I,I 30 | NP_001307731.1 | |
NM_001320803.1 | 125 | Silent Mutation | ATC,ATT | I,I 30 | NP_001307732.1 | |
NM_139280.3 | 125 | Silent Mutation | ATC,ATT | I,I 30 | NP_644809.1 | |
XM_017025345.1 | 125 | Silent Mutation | ATC,ATT | I,I 48 | XP_016880834.1 | |
XM_017025346.1 | 125 | Silent Mutation | ATC,ATT | I,I 30 | XP_016880835.1 | |
XM_017025347.1 | 125 | Silent Mutation | ATC,ATT | I,I 30 | XP_016880836.1 | |
XM_017025348.1 | 125 | Silent Mutation | ATC,ATT | I,I 30 | XP_016880837.1 |