Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTCCAAGCCTGGCATGGACCTG[A/C]AGAGACCCGATTCCTACCAGGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609956 | ||||||||||||||||||||
Literature Links: |
RAB37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB37 - RAB37, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006638.2 | 480 | Intron | NP_001006639.1 | |||
NM_001163989.1 | 480 | Intron | NP_001157461.1 | |||
NM_001163990.1 | 480 | Intron | NP_001157462.1 | |||
NM_175738.4 | 480 | Missense Mutation | AAG,CAG | K,Q 4 | NP_783865.1 | |
XM_005257288.4 | 480 | Intron | XP_005257345.1 |