Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGCACACTTCACCCATCCCCT[G/T]TTGTGGGCCATTTAGGTTGCTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LLGL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LLGL2 - LLGL2, scribble cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015002.1 | Intron | NP_001015002.1 | ||||
NM_001031803.1 | Intron | NP_001026973.1 | ||||
NM_004524.2 | Intron | NP_004515.2 | ||||
XM_011524801.1 | Intron | XP_011523103.1 | ||||
XM_011524802.1 | Intron | XP_011523104.1 | ||||
XM_011524807.2 | Intron | XP_011523109.1 | ||||
XM_017024625.1 | Intron | XP_016880114.1 | ||||
XM_017024626.1 | Intron | XP_016880115.1 | ||||
XM_017024627.1 | Intron | XP_016880116.1 | ||||
XM_017024628.1 | Intron | XP_016880117.1 | ||||
XM_017024629.1 | Intron | XP_016880118.1 | ||||
XM_017024630.1 | Intron | XP_016880119.1 | ||||
XM_017024631.1 | Intron | XP_016880120.1 | ||||
XM_017024632.1 | Intron | XP_016880121.1 |