Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCACGCAGCGGCAGGAAGGCCTC[A/G]CCTTCCAGGTCGTCGGCCCCCAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608897 MIM: 616375 | ||||||||||||||||||||
Literature Links: |
LOC100507191 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507191 - uncharacterized LOC100507191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC13D - unc-13 homolog D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199242.2 | 3127 | Silent Mutation | GGC,GGT | G,G 1016 | NP_954712.1 | |
XM_011524504.1 | 3127 | Silent Mutation | GGC,GGT | G,G 1039 | XP_011522806.1 | |
XM_011524506.1 | 3127 | Silent Mutation | GGC,GGT | G,G 1038 | XP_011522808.1 | |
XM_011524507.2 | 3127 | Silent Mutation | GGC,GGT | G,G 836 | XP_011522809.1 |
UNK - unkempt family zinc finger | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |