Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAATAAAAAGTCGTTTTAGTC[A/C]ATTACAGTGTGACTTGTTCTAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609751 MIM: 616807 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOX1 - acyl-CoA oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185039.1 | 6594 | UTR 3 | NP_001171968.1 | |||
NM_004035.6 | 6594 | UTR 3 | NP_004026.2 | |||
NM_007292.5 | 6594 | UTR 3 | NP_009223.2 | |||
XM_011524868.2 | 6594 | UTR 3 | XP_011523170.1 | |||
XM_011524869.2 | 6594 | UTR 3 | XP_011523171.1 |
FBF1 - Fas binding factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |