Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGATGTAGGGCAGGAAGGCGAA[A/G]GCCAGCAGCAGGCCACTGAGGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600950 MIM: 614404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AANAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AANAT - aralkylamine N-acetyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDF2 - rhomboid 5 homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005498.3 | 3007 | Silent Mutation | GCC,GCT | A,A 755 | NP_001005498.2 | |
NM_024599.5 | 3007 | Silent Mutation | GCC,GCT | A,A 784 | NP_078875.4 | |
XM_005257669.3 | 3007 | Silent Mutation | GCC,GCT | A,A 784 | XP_005257726.1 | |
XM_005257670.1 | 3007 | Silent Mutation | GCC,GCT | A,A 755 | XP_005257727.1 | |
XM_011525249.2 | 3007 | Silent Mutation | GCC,GCT | A,A 755 | XP_011523551.1 | |
XM_011525250.2 | 3007 | Silent Mutation | GCC,GCT | A,A 755 | XP_011523552.1 | |
XM_011525251.2 | 3007 | Silent Mutation | GCC,GCT | A,A 755 | XP_011523553.1 | |
XM_017025079.1 | 3007 | Silent Mutation | GCC,GCT | A,A 755 | XP_016880568.1 |