Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAATAACAGCTGGAACCTCACCA[A/G]CAAGCAGAAACCAGAGGGGTCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105274304 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105274304 - uncharacterized LOC105274304 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MXRA7 - matrix remodeling associated 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008528.2 | 903 | UTR 3 | NP_001008528.1 | |||
NM_001008529.2 | 903 | Intron | NP_001008529.1 | |||
NM_198530.3 | 903 | Intron | NP_940932.2 | |||
XM_005257382.2 | 903 | UTR 3 | XP_005257439.1 | |||
XM_005257383.2 | 903 | Intron | XP_005257440.1 | |||
XM_005257384.2 | 903 | Intron | XP_005257441.1 | |||
XM_011524830.1 | 903 | UTR 3 | XP_011523132.1 |