Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATCTCTCCTCCCCTGTCAAACC[A/G]GTGACAGAAGTGGGGTCCCACATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612441 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGAT5B - mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199172.1 | Intron | NP_001186101.1 | ||||
NM_144677.2 | Intron | NP_653278.2 | ||||
NM_198955.1 | Intron | NP_945193.1 | ||||
XM_006721707.3 | Intron | XP_006721770.1 | ||||
XM_011524350.2 | Intron | XP_011522652.1 | ||||
XM_011524352.2 | Intron | XP_011522654.1 | ||||
XM_011524354.2 | Intron | XP_011522656.1 | ||||
XM_017024200.1 | Intron | XP_016879689.1 |