Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTAACATTCAGTCAAAAACTAA[A/G]CATATAAAAAGAGGCAGGCGATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNRC6C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNRC6C - trinucleotide repeat containing 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142640.1 | 4034 | Intron | NP_001136112.1 | |||
NM_018996.3 | 4034 | Intron | NP_061869.2 | |||
XM_006721996.3 | 4034 | UTR 5 | XP_006722059.3 | |||
XM_006721997.3 | 4034 | UTR 5 | XP_006722060.3 | |||
XM_006722000.3 | 4034 | UTR 5 | XP_006722063.3 | |||
XM_011525088.2 | 4034 | Intron | XP_011523390.1 | |||
XM_017024906.1 | 4034 | UTR 5 | XP_016880395.1 | |||
XM_017024907.1 | 4034 | UTR 5 | XP_016880396.1 | |||
XM_017024908.1 | 4034 | UTR 5 | XP_016880397.1 |