Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAGCCTCTTAAAGCTCCACTCC[A/G]CCAAATGCAGTTGACTACCTTTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371910 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371910 - uncharacterized LOC105371910 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM235 - transmembrane protein 235 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204210.1 | 1470 | Intron | NP_001191139.1 | |||
NM_001204211.1 | 1470 | Intron | NP_001191140.1 | |||
NM_001204212.1 | 1470 | Intron | NP_001191141.1 | |||
XM_011524620.2 | 1470 | Intron | XP_011522922.1 | |||
XM_011524624.2 | 1470 | Intron | XP_011522926.1 | |||
XM_017024459.1 | 1470 | Intron | XP_016879948.1 | |||
XM_017024460.1 | 1470 | Intron | XP_016879949.1 | |||
XM_017024461.1 | 1470 | UTR 3 | XP_016879950.1 |