Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCTGCTTTGCAGACATAACAG[C/T]TGGGAGCCGGAGGAGAACATCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBX2 - chromobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005189.2 | 184 | Silent Mutation | AGC,AGT | S,S 42 | NP_005180.1 | |
NM_032647.3 | 184 | Silent Mutation | AGC,AGT | S,S 42 | NP_116036.1 | |
XM_011525382.1 | 184 | Silent Mutation | AGC,AGT | S,S 42 | XP_011523684.1 | |
XM_011525383.2 | 184 | UTR 5 | XP_011523685.1 |