Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGTCGTGGCTCTGGTTGCAGCA[C/G]CTCAGACGAGTGCGGGACCCGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPTOR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPTOR - regulatory associated protein of MTOR complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163034.1 | 280 | UTR 5 | NP_001156506.1 | |||
NM_020761.2 | 280 | UTR 5 | NP_065812.1 |