Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAGCACTGGCAGCTTCACTGAC[C/T]TGTAGATCATCTGCAGACCCCAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608588 | ||||||||||||||||||||
Literature Links: |
DHX58 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX58 - DEXH-box helicase 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024119.2 | 2096 | Missense Mutation | AAG,AGG | K,R 626 | NP_077024.2 | |
XM_017025059.1 | 2096 | Missense Mutation | AAG,AGG | K,R 626 | XP_016880548.1 | |
XM_017025060.1 | 2096 | Missense Mutation | AAG,AGG | K,R 626 | XP_016880549.1 |