Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACACTGTTCCACCACCAGGCTGA[G/T]GCTGGGTGGCCGGCTTCCCAACCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608344 MIM: 602301 MIM: 604037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPB9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPB9 - heat shock protein family B (small) member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KAT2A - lysine acetyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB5C - RAB5C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252039.1 | Intron | NP_001238968.1 | ||||
NM_004583.3 | Intron | NP_004574.2 | ||||
NM_201434.2 | Intron | NP_958842.1 | ||||
XM_011525090.1 | Intron | XP_011523392.1 | ||||
XM_011525091.1 | Intron | XP_011523393.1 |