Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCTTTCCCTCTTAGCCACCAC[A/G]GGCAGTGACCCCAGTTTGTCTCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608463 MIM: 601999 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AATF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AATF - apoptosis antagonizing transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LHX1 - LIM homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005568.4 | 1128 | Silent Mutation | ACA,ACG | T,T 135 | NP_005559.2 |
LOC102723471 - uncharacterized LOC102723471 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |