Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCACATCAAGCAGCTTTACCT[G/T]CAGGTAAAAGGATGGAAAAGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAGLU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAGLU - N-acetyl-alpha-glucosaminidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000263.3 | 752 | Silent Mutation | CTG,CTT | L,L 225 | NP_000254.2 | |
XM_006721920.2 | 752 | UTR 5 | XP_006721983.1 | |||
XM_011524840.1 | 752 | UTR 5 | XP_011523142.1 | |||
XM_017024686.1 | 752 | Silent Mutation | CTG,CTT | L,L 22 | XP_016880175.1 | |
XM_017024687.1 | 752 | UTR 5 | XP_016880176.1 |