Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCGATCTCCCTAGGCCTGATTTT[C/T]ACGGCCCCTTTGTGGCCCTGAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610907 MIM: 601844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS25 - vacuolar protein sorting 25 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032353.3 | Intron | NP_115729.1 | ||||
XM_017025224.1 | Intron | XP_016880713.1 |
WNK4 - WNK lysine deficient protein kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |