Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGCCAGGATGACAGCAGTTCC[A/G]AAGATGCCCACACTCTCTCCAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608300 MIM: 600781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAGS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAGS - N-acetylglutamate synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYY - peptide YY | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004160.5 | 986 | Intron | NP_004151.3 | |||
XM_011525035.1 | 986 | Intron | XP_011523337.1 |
TMEM101 - transmembrane protein 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304813.1 | 986 | Silent Mutation | TTC,TTT | F,F 189 | NP_001291742.1 | |
NM_001304814.1 | 986 | Silent Mutation | TTC,TTT | F,F 189 | NP_001291743.1 | |
NM_032376.3 | 986 | Silent Mutation | TTC,TTT | F,F 247 | NP_115752.1 | |
XM_005257737.2 | 986 | Silent Mutation | TTC,TTT | F,F 247 | XP_005257794.1 | |
XM_011525346.1 | 986 | Silent Mutation | TTC,TTT | F,F 247 | XP_011523648.1 | |
XM_011525347.1 | 986 | Silent Mutation | TTC,TTT | F,F 247 | XP_011523649.1 | |
XM_011525349.1 | 986 | Silent Mutation | TTC,TTT | F,F 247 | XP_011523651.1 | |
XM_011525351.1 | 986 | Silent Mutation | TTC,TTT | F,F 247 | XP_011523653.1 | |
XM_011525353.1 | 986 | Silent Mutation | TTC,TTT | F,F 189 | XP_011523655.1 |