Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTTGTGAGGATAAATTCCATTC[A/G]TTAGGGCAGACACAGATCACAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBF4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBF4B - DBF4 zinc finger B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025104.3 | Intron | NP_079380.2 | ||||
NM_145663.2 | Intron | NP_663696.1 | ||||
XM_006722098.2 | Intron | XP_006722161.1 | ||||
XM_006722102.3 | Intron | XP_006722165.1 | ||||
XM_011525281.2 | Intron | XP_011523583.1 | ||||
XM_017025145.1 | Intron | XP_016880634.1 | ||||
XM_017025146.1 | Intron | XP_016880635.1 | ||||
XM_017025147.1 | Intron | XP_016880636.1 | ||||
XM_017025148.1 | Intron | XP_016880637.1 | ||||
XM_017025149.1 | Intron | XP_016880638.1 | ||||
XM_017025150.1 | Intron | XP_016880639.1 | ||||
XM_017025151.1 | Intron | XP_016880640.1 | ||||
XM_017025152.1 | Intron | XP_016880641.1 | ||||
XM_017025153.1 | Intron | XP_016880642.1 | ||||
XM_017025154.1 | Intron | XP_016880643.1 | ||||
XM_017025155.1 | Intron | XP_016880644.1 | ||||
XM_017025156.1 | Intron | XP_016880645.1 |