Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611519 MIM: 607732 MIM: 610975 MIM: 616815 MIM: 193190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4723 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4723 - microRNA 4723 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLDIP2 - DNA polymerase delta interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SARM1 - sterile alpha and TIR motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEBOX - SEBOX homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080837.3 | 211 | Missense Mutation | CGG,TGG | R,W 71 | NP_001074306.3 |
TMEM199 - transmembrane protein 199 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152464.2 | 211 | Intron | NP_689677.1 |
VTN - vitronectin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |